Learn More
Selleck Chemical LLC Selenomethionine
SDP
Catalog No. 5000004064
Encompass_Preferred
Selenomethionine (Seleno-DL-methionine DL-Selenomethionine) is a naturally occurring amino acid containing selenium Selenomethionine (SeMet) can substitute for methionine (Met) during protein synthesis in a non-specific manner Due to the significant differences in their redox properties the incorporation of SeMet in place of Met may adversely impact protein function
Catalog No. 50-000-04064 Supplier Selleck Chemical LLC Supplier No. S597050MG
Selenomethionine (Seleno-DL-methionine DL-Selenomethionine) is a naturally occurring amino acid containing selenium Selenomethionine (SeMet) can substitute for methionine (Met) during protein synthesis in a non-specific manner Due to the significant differences in their redox properties the incorporation of SeMet in place of Met may adversely impact protein function