Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selenomethionine (Standard) | 1464-42-2 | 99.4% | 25 MG
SDP

Catalog No. 5000450878
Encompass_Preferred

Selenomethionine (Standard) is the analytical standard of Selenomethionine, a naturally occurring amino acid containing selenium. Intended for research and analytical applications, it exhibits oral activity and has demonstrated antitumor activity. This compound is suitable for qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.

  • Naturally occurring amino acid
  • Contains selenium
  • Oral activity demonstrated
  • Common natural food source
  • Exhibits antitumor activity
  • Suitable for HPLC, GC, and MS research
  • Molecular weight: 196.11
  • Chemical formula: C5H11NO2Se
  • Appearance: solid
  • Color: white to off-white
  • Classified as an amino acid
  • Functions as an endogenous metabolite
  • Disease marker for cancer and nervous system disorders
  • Found in microorganisms

Catalog No. 50-004-50878 Supplier Medchemexpress LLC Supplier No. 25MGHYB1000R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selenomethionine (Standard) is the analytical standard of Selenomethionine, a naturally occurring amino acid containing selenium. Intended for research and analytical applications, it exhibits oral activity and has demonstrated antitumor activity. This compound is suitable for qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.

  • Naturally occurring amino acid
  • Contains selenium
  • Oral activity demonstrated
  • Common natural food source
  • Exhibits antitumor activity
  • Suitable for HPLC, GC, and MS research
  • Molecular weight: 196.11
  • Chemical formula: C5H11NO2Se
  • Appearance: solid
  • Color: white to off-white
  • Classified as an amino acid
  • Functions as an endogenous metabolite
  • Disease marker for cancer and nervous system disorders
  • Found in microorganisms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.