Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selenomethionine (Standard) | 1464-42-2 | 99.4% | 50 MG
SDP

Catalog No. 5000407506
Encompass_Preferred

Selenomethionine (Standard) is an analytical standard of Selenomethionine, intended for research and analytical applications. It is a naturally occurring amino acid containing selenium that has oral activity and antitumor activity.

  • Analytical standard for research and analytical applications
  • Naturally occurring amino acid containing selenium
  • Oral activity
  • Antitumor activity
  • Molecular formula: C5H11NO2Se
  • Molecular weight: 196.11

Catalog No. 50-004-07506 Supplier Medchemexpress LLC Supplier No. HYB1000R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selenomethionine (Standard) is an analytical standard of Selenomethionine, intended for research and analytical applications. It is a naturally occurring amino acid containing selenium that has oral activity and antitumor activity.

  • Analytical standard for research and analytical applications
  • Naturally occurring amino acid containing selenium
  • Oral activity
  • Antitumor activity
  • Molecular formula: C5H11NO2Se
  • Molecular weight: 196.11

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.