Learn More
Medchemexpress LLC Selenomethionine (standard) | 1464-42-2 | MFCD00063089 | 99.4% | 196.11 | C5H11NO2Se | 10 MG
SDP
Catalog No. 5000297732
Encompass_Preferred
Selenomethionine (standard) is an analytical reference standard of selenomethionine supplied as a 10 mg pack for research and analytical applications. It is used as a reference material for qualitative and quantitative methods and is suitable for HPLC, GC, and MS analyses. The material has a reported purity of 99.4% and a molecular weight of 196.11.
  • Analytical reference standard for selenomethionine.
  • Intended for qualitative and quantitative HPLC, GC, and MS assays.
  • High purity (99.4%) for reliable quantitation.
  • Compact 10 mg pack size for analytical use.
  • CAS number 1464-42-2; molecular weight 196.11.
Catalog No. 50-002-97732 Supplier Medchemexpress LLC Supplier No. HYB1000R10MG
Selenomethionine (standard) is an analytical reference standard of selenomethionine supplied as a 10 mg pack for research and analytical applications. It is used as a reference material for qualitative and quantitative methods and is suitable for HPLC, GC, and MS analyses. The material has a reported purity of 99.4% and a molecular weight of 196.11.
  • Analytical reference standard for selenomethionine.
  • Intended for qualitative and quantitative HPLC, GC, and MS assays.
  • High purity (99.4%) for reliable quantitation.
  • Compact 10 mg pack size for analytical use.
  • CAS number 1464-42-2; molecular weight 196.11.