Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selenomethylene blue | 85670-01-5 | 96.2% | 366.75 g/mol | C16H18ClN3Se | 10 MG
SDP

Supplier:  Medchemexpress LLC HYD178210MG

Encompass_Preferred

Selenomethylene blue is a selenium-containing dye used as an antioxidant and anti-inflammatory research reagent. It has been reported to inhibit inflammatory paw edema in animal models and is applied as a selective selenol fluorescent probe for imaging and quantifying selenoprotein activity.

  • Acts as an antioxidant and anti-inflammatory agent.
  • Functions as a selective selenol fluorescent probe for imaging and quantification.
  • CAS number 85670-01-5.
  • Molecular formula C16H18ClN3Se and molecular weight 366.75 g/mol.
  • Purity approximately 96.2%.
  • Available in small research quantities including 10 mg.

Catalog No. 50-002-97189


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.