Learn More
Medchemexpress LLC Selenosemicarbazide | 21198-79-8 | MFCD00216963 | 95.0% | 138.03 | CH5N3Se | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1282225MG
Encompass_Preferred
Selenosemicarbazide (CH5N3Se, CAS 21198-79-8) is a selenium-containing small molecule used as a research reagent. It has documented antimicrobial activity and can form metal complexes that have been investigated for antitumor effects in cell studies. The substance is supplied as a small-quantity solid suitable for analytical, synthetic, and biological research; intended for laboratory research use only.
  • Exhibits antimicrobial activity against a range of bacteria.
  • Forms metal complexes with reported antitumor activity in cell-based studies.
  • Serves as a reagent or building block in organoselenium research and synthesis.
  • Provided in small-quantity solid formats suitable for analytical and synthetic workflows.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-63425