Learn More
Selenous acid, 97%
Description
As an oxidizing agent, as a reagent for alkaloidsSelenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Melting Point | 70°C (decomposition) |
| Density | 3.004g/cm3 at 20°C |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Assay Percent Range | 97% |
| Quantity | 50 g |
| Physical Form | -4 Mesh Crystalline |
| Solubility Information | Freely soluble in water,alcohol. Insoluble in ammoniaSoluble in water and ethanol. |
| Formula Weight | 128.97 |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H373 |
| Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: II |
| EINECSNumber | 231-974-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.