Learn More
Medchemexpress LLC Selepressin acetate | 00-00-0 | 98.8% | 1048.28 g/mol (free base) | C48H77N13O13S2 | 10 MG
SDP
Catalog No. 5000243926
Encompass_Preferred
Selepressin acetate is a synthetic peptide vasopressin V1A receptor agonist used in preclinical research to study septic shock and vascular barrier function. It acts as a potent vasopressor and has demonstrated efficacy in reducing vascular leak and preserving endothelial integrity in cell and animal models.
  • Selective vasopressin V1A receptor agonist.
  • Potent vasopressor that increases mean arterial pressure in endotoxemia models.
  • Reduces vascular leak and protects endothelial barrier in vitro and in vivo.
  • High purity solid peptide suitable for research applications.
  • Stable when stored frozen; follow supplier storage recommendations.
Catalog No. 50-002-43926 Supplier Medchemexpress LLC Supplier No. HY105239A10MG
Selepressin acetate is a synthetic peptide vasopressin V1A receptor agonist used in preclinical research to study septic shock and vascular barrier function. It acts as a potent vasopressor and has demonstrated efficacy in reducing vascular leak and preserving endothelial integrity in cell and animal models.
  • Selective vasopressin V1A receptor agonist.
  • Potent vasopressor that increases mean arterial pressure in endotoxemia models.
  • Reduces vascular leak and protects endothelial barrier in vitro and in vivo.
  • High purity solid peptide suitable for research applications.
  • Stable when stored frozen; follow supplier storage recommendations.