Learn More
Medchemexpress LLC Selepressin acetate 1mg | 1MG
SDP
Catalog No. 5000242401
Encompass_Preferred
Selepressin (FE 202158) acetate is a selective vasopressin V1A receptor agonist Selepressin acetate is a potent vasopressin Selepressin acetate can be used in the study of septic shock
Catalog No. 50-002-42401 Supplier Medchemexpress LLC Supplier No. HY105239A1MG
Selepressin (FE 202158) acetate is a selective vasopressin V1A receptor agonist Selepressin acetate is a potent vasopressin Selepressin acetate can be used in the study of septic shock