Learn More
Medchemexpress LLC Selexipag | 475086-01-2 | 496.62 | 100 MG
SDP
Catalog No. 5000362425
Encompass_Preferred
Selexipag (NS-304) is an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor). It is also described as an orally available and long-acting IP receptor agonist prodrug, with its active form being MRE-269.
  • Orally available and potent agonist for the prostacyclin (PGI2) receptor (IP receptor)
  • Long-acting IP receptor agonist prodrug; active form is MRE-269
  • Inhibits binding of [3H]Iloprost to human and rat IP receptors
  • Increases intracellular cAMP levels in hIP-CHO cells
  • Inhibits platelet aggregation in humans and monkeys
Catalog No. 50-003-62425 Supplier Medchemexpress LLC Supplier No. HY14870100MG
Selexipag (NS-304) is an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor). It is also described as an orally available and long-acting IP receptor agonist prodrug, with its active form being MRE-269.
  • Orally available and potent agonist for the prostacyclin (PGI2) receptor (IP receptor)
  • Long-acting IP receptor agonist prodrug; active form is MRE-269
  • Inhibits binding of [3H]Iloprost to human and rat IP receptors
  • Increases intracellular cAMP levels in hIP-CHO cells
  • Inhibits platelet aggregation in humans and monkeys