Learn More
Medchemexpress LLC Selexipag-d7 | 1265295-21-3 | 98.4% | 503.66 | 1 MG
SDP
Catalog No. 5000361393
Encompass_Preferred
Selexipag-d7 is the deuterium-labeled Selexipag, an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor). Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers in drug development, with potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-003-61393 Supplier Medchemexpress LLC Supplier No. HY14870S11MG
Selexipag-d7 is the deuterium-labeled Selexipag, an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor). Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers in drug development, with potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only