Learn More
MSC Justrite Flammable Cabinet, 45 gal., Self Closing, Yellow, 1 ea
Catalog No. 502886288
Encompass
This Justrite Flammable and Hazardous Storage Cabinet is designed for the safe storage of flammable and combustible liquids. Featuring a 45 gal. capacity, two self-closing doors, and two shelves, it provides secure containment and compliance in laboratory and industrial environments.
  • Equipped with a 3-step self-close system, activating upon fire via a fusible link at 165°F (74°C)
  • Doors spring shut with inertia and latches engage for secure closure
  • Two adjustable shelves for versatile storage configurations
  • Features a 3 point key lock for enhanced security
  • Floor mount design for stable placement
  • Constructed from durable steel for robust protection
  • Compliant with NFPA 1, FM, OSHA, NFPA 30, and IFC safety standards
Catalog No. 50-288-6288 Supplier MSC Supplier No. 89847073
This Justrite Flammable and Hazardous Storage Cabinet is designed for the safe storage of flammable and combustible liquids. Featuring a 45 gal. capacity, two self-closing doors, and two shelves, it provides secure containment and compliance in laboratory and industrial environments.
  • Equipped with a 3-step self-close system, activating upon fire via a fusible link at 165°F (74°C)
  • Doors spring shut with inertia and latches engage for secure closure
  • Two adjustable shelves for versatile storage configurations
  • Features a 3 point key lock for enhanced security
  • Floor mount design for stable placement
  • Constructed from durable steel for robust protection
  • Compliant with NFPA 1, FM, OSHA, NFPA 30, and IFC safety standards