Learn More
SPS Medical SELF CONTAIN STEAM BIVIAL25/PK
Catalog No. NC9974089
Encompass_Preferred
The SporView Self-Contained Biological (BIAEs) are approved for use in gravity, prevacuum, and flash cycles. SporViewPlus can also be purchased with 50 BIs and 50 STEAMPlus integrators per pack, for immediate release of routine or flashed loads. Final visual spore growth is recorded in 24 hours. For new customers, a starter kit is available with an incubator, record notebook and 25 BIs.
Catalog No. NC9974089 Supplier SPS Medical Supplier No. SCS025
The SporView Self-Contained Biological (BIAEs) are approved for use in gravity, prevacuum, and flash cycles. SporViewPlus can also be purchased with 50 BIs and 50 STEAMPlus integrators per pack, for immediate release of routine or flashed loads. Final visual spore growth is recorded in 24 hours. For new customers, a starter kit is available with an incubator, record notebook and 25 BIs.