Learn More
HW ANDERSEN PRODUCTS OF CALIFORNIA INC Biological Indicator EZ Test, 25/box
SDP
Catalog No. NC1419451
Encompass
Self-contained Biological Indicators (BI’s) provide sterility assurance for EtO cycles. High population of spores (106 B. athrophaeus) achieves accuracy. Color change reveals EtO test results after 48 hours incubation at 37°C.
Catalog No. NC1419451 Supplier HW ANDERSEN PRODUCTS OF CALIFORNIA INC Supplier No. AN2203.00
Self-contained Biological Indicators (BI’s) provide sterility assurance for EtO cycles. High population of spores (106 B. athrophaeus) achieves accuracy. Color change reveals EtO test results after 48 hours incubation at 37°C.