Learn More
MSC OccuNomix High Visibility Safety Vest, Polyester Mesh, Self-Extinguishing, ANSI Type R Class 2, 1 ea
Catalog No. 5000688859
Encompass
This high-visibility safety vest is designed for professionals requiring enhanced visibility and flame-resistant properties. Constructed from polyester mesh, it meets ANSI/ISEA 107-2020 Type R Class 2 standards for workplace safety. The material is treated to self-extinguish and remains ASTM D6413 compliant for up to 25 washes.
  • Meets ANSI/ISEA 107-2020 Type R Class 2 requirements
  • Treated to self-extinguish
  • Compliant with ASTM D6413 for up to 25 washes
  • Features 2 in. flame-resistant reflective tape
  • Equipped with a quick-release zipper
  • Includes two pockets for storage, consisting of one left chest pocket and one inner lower pocket
Catalog No. 50-006-88859 Supplier MSC Supplier No. 19326248
This high-visibility safety vest is designed for professionals requiring enhanced visibility and flame-resistant properties. Constructed from polyester mesh, it meets ANSI/ISEA 107-2020 Type R Class 2 standards for workplace safety. The material is treated to self-extinguish and remains ASTM D6413 compliant for up to 25 washes.
  • Meets ANSI/ISEA 107-2020 Type R Class 2 requirements
  • Treated to self-extinguish
  • Compliant with ASTM D6413 for up to 25 washes
  • Features 2 in. flame-resistant reflective tape
  • Equipped with a quick-release zipper
  • Includes two pockets for storage, consisting of one left chest pocket and one inner lower pocket