Learn More
MSC OccuNomix Flame Resistant Safety Apparel, Polyester Mesh, Orange, XL, 1/Each
Catalog No. 5000688874
Encompass
This high-visibility safety vest is constructed from polyester mesh treated to self-extinguish, designed for use in environments where flame resistance is required, such as electrical and utility work. The garment features a quick-release zipper for safety and convenience.
  • Meets ANSI/ISEA 107-2020 Type R Class 2 compliance
  • Treated to self-extinguish and ASTM D6413 compliant after 25 washes
  • Equipped with 2 in. flame-resistant reflective tape
  • Includes a left chest pocket and an inner lower pocket
  • Quick-release zipper design
Catalog No. 50-006-88874 Supplier MSC Supplier No. 19326255
This high-visibility safety vest is constructed from polyester mesh treated to self-extinguish, designed for use in environments where flame resistance is required, such as electrical and utility work. The garment features a quick-release zipper for safety and convenience.
  • Meets ANSI/ISEA 107-2020 Type R Class 2 compliance
  • Treated to self-extinguish and ASTM D6413 compliant after 25 washes
  • Equipped with 2 in. flame-resistant reflective tape
  • Includes a left chest pocket and an inner lower pocket
  • Quick-release zipper design