Learn More
Waters Corp Flush Assembly, Automatic, Seal Wash, Reduces Seal Wear
Catalog No. 50792325 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This flush assembly is designed to reduce seal wear and prevent crystal deposition in pump heads during operation. It continuously washes the region behind the primary high-pressure seal, ensuring optimal pump performance and longevity.
  • Continuously washes the pump head behind the primary high-pressure seal during operation.
  • Reduces seal wear.
  • Prevents crystal deposition from buffered solvents.
  • Helps avoid premature seal failure, leakage, and pump damage.
  • Uses a seal wash reservoir and seal wash inlet/outlet tubing.
  • Requires a seal wash solvent.
Catalog No. 50-792-325 Supplier Waters Corp Supplier No. 700000179
This flush assembly is designed to reduce seal wear and prevent crystal deposition in pump heads during operation. It continuously washes the region behind the primary high-pressure seal, ensuring optimal pump performance and longevity.
  • Continuously washes the pump head behind the primary high-pressure seal during operation.
  • Reduces seal wear.
  • Prevents crystal deposition from buffered solvents.
  • Helps avoid premature seal failure, leakage, and pump damage.
  • Uses a seal wash reservoir and seal wash inlet/outlet tubing.
  • Requires a seal wash solvent.