Learn More
GA International Self-Laminating Thermal-Transfer Labels for Microscope Slides 0.88" x 0.97" / 22.4mm x 24.6mm Rectangle White From -80°C to +120°C (-112°F to 248°F)
Catalog No. 5000572510
Encompass_Preferred
Xylene-resistant, thermal-transfer labels specially designed for slide identification in fully automated, multi-slide, immunohistochemistry (IHC), and special stainers and processors.Resistant to routinely used slide/tissue processing and immunohistochemical staining harsh chemicals such as Xylene, ethanol, formalin, acetic acid, as well as eosin, hematoxylin, and special (counter) stains. Specially designed with an over-laminating flap, these labels can also withstand long-term exposure and immersion in safer clearing and deparaffinizing Xylene substitutes such as EZ Prep, Clearify, Histo-Clear, Formula 83, Pro-Par, and others. Resistant to high-temperature, acidic/basic buffers used during the antigen retrieval process in immunohistochemistry protocols.Capable of integrating with laboratory information systems (LIS) and management systems (LIMS), as well as slide processors’ specific label printers. They are ideal for safe, accurate, and permanent identification of microscope sli
Catalog No. 50-005-72510 Supplier GA International Supplier No. AFT1C31WH
Xylene-resistant, thermal-transfer labels specially designed for slide identification in fully automated, multi-slide, immunohistochemistry (IHC), and special stainers and processors.Resistant to routinely used slide/tissue processing and immunohistochemical staining harsh chemicals such as Xylene, ethanol, formalin, acetic acid, as well as eosin, hematoxylin, and special (counter) stains. Specially designed with an over-laminating flap, these labels can also withstand long-term exposure and immersion in safer clearing and deparaffinizing Xylene substitutes such as EZ Prep, Clearify, Histo-Clear, Formula 83, Pro-Par, and others. Resistant to high-temperature, acidic/basic buffers used during the antigen retrieval process in immunohistochemistry protocols.Capable of integrating with laboratory information systems (LIS) and management systems (LIMS), as well as slide processors’ specific label printers. They are ideal for safe, accurate, and permanent identification of microscope sli