Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Self-laminating Vinyl Wire and Cable Labels

Catalog No. 19129808 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
1.5 in. H x 1.5 in. W
Quantity:
180 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Quantity
19-129-808 1.5 in. H x 1.5 in. W 180 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-808 Supplier Brady™ Supplier No. M32427

Brady™ Self-laminating Vinyl Wire and Cable Labels (B-427) are excellent for wire and cable identification. The material has good clarity, conformability, and is self-extinguishing.

  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
  • Durable, permanent acrylic adhesive with water and oil resistance
  • UL approved
  • All-in-one cartridge includes ribbon and label supply

Specifications

Product Type Label
Color Black on White/Transparent
For Use With (Equipment) BMP51 Label Printer, BMP53 Label Printer
Material Self-laminating Vinyl (B-427)
Print Technology Thermal Transfer
Dimensions (L x W) 1.5 in. H x 1.5 in. W
Adhesive Type Permanent Acrylic
Sterility Non-sterile
Finish Matte
Format Cartridge, Ribbon
Certifications/Compliance UL Approved; Compliant with Directive 2011/65/EU and 2015/863
Quantity 180 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.