Learn More
Brady™ Self-Laminating Vinyl Wrap Around Labels 5000/Roll
Catalog No. 19221546 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Gray
Green
Orange
Pink
Purple
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-221-546 Orange
19-221-492 Green
19-221-520 Gray
19-221-522 Pink
19-221-466 Purple
19-221-503 Red
19-221-408 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-221-546 Supplier Brady™ Supplier No. B338417OR
Add to Cart
Add to Cart
Secure your wire identification with high-performance material and extra-strong adhesive.
  • Features a white or color printable area and a clear tail that wraps around the the marker and protects the print
  • Extra strength adhesive for standard wire and cable identification and also adhesion in high-humidity conditions, on smaller diameter wires, and on challenging wire jacket materials including Teflon™, silicone, braided and traditional PVC
  • Self-extinguishing material with outstanding resistance to abrasion, handling, oil, water and dirt
  • Select sizes available in 12 colors for TIA-606-B and TIA 598 color coding of data center cabling, network infrastructure, and electrical wires and cables
  • A ribbon is required to print this material. Use Part Number B30-R4300
Specifications
Adhesive Type Permanent Acrylic
Color Orange
Finish Matte
Format Label
Material Self-laminating Vinyl
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Length (English) 1.437 in.
Width (English) 0.5 in.