Learn More
World Precision Instrument Self-Retaining Retractor, 7.5cm, Square Blade
SDP
Catalog No. 502539372
Encompass
  • 7.5cm (3") Long
  • 1.2cm Square blade
  • Maximum spread: 3cm
Catalog No. 50-253-9372 Supplier World Precision Instrument Supplier No. 504472
  • 7.5cm (3") Long
  • 1.2cm Square blade
  • Maximum spread: 3cm