Learn More
Andwin Scientific SELF SEAL POUCH 71/2X13 200/PK
SDP
Catalog No. NC9894917
Encompass_Preferred
V. Mueller DualPeel Self-Sealing Pouch, Pressure-sensitive closure is prefolded and perforated to assure seal integrity. Blue-tinted viewing area. High porosity for fast sterilization while maintaining effective bacterial barrier.
Catalog No. NC9894917 Supplier Andwin Scientific Supplier No. 92713PK
V. Mueller DualPeel Self-Sealing Pouch, Pressure-sensitive closure is prefolded and perforated to assure seal integrity. Blue-tinted viewing area. High porosity for fast sterilization while maintaining effective bacterial barrier.