Learn More
CEM Corporation SELF SEALING QUARTZ CRUCIBLE
SDP
Supplier:  CEM Corporation 322200
Encompass
Self Sealing Quartz Crucibles, Set of 4. Includes crucibles and lids for oxygen free ashing
Catalog No. NC0953892