Learn More
Analytical Sales & Services Self Standing 1.8mL Low Adhesion Screw Top Micro Tube
Catalog No. 5000804569
Encompass
Self Standing 1.8mL Low Adhesion Screw Top Micro Tube\nCS 1000, Polypropylene\nTemperature Range: -196 ¡C to +121 ¡C\nCertified RNase-, DNase-, Pyrogen- and DNA-free.\n\nQty/Pkg: 1000
Catalog No. 50-008-04569 Supplier Analytical Sales & Services Supplier No. 16603
Self Standing 1.8mL Low Adhesion Screw Top Micro Tube\nCS 1000, Polypropylene\nTemperature Range: -196 ¡C to +121 ¡C\nCertified RNase-, DNase-, Pyrogen- and DNA-free.\n\nQty/Pkg: 1000