Learn More
Alkali Scientific Self Standing Transport Tubes, 10mL, with Separate Screw Cap, AMBER, PP, 1000/Cs
SDP
Catalog No. 5000663188
Encompass_Preferred
Transport tubes and vials are designed for the safe storage and transport of biological materials in laboratory and research environments. Manufactured from transparent polypropylene (PP), these vials allow clear and unobstructed viewing of the sample during handling and inspection. The durable polypropylene construction provides excellent resistance to breakage, making the vials shatter proof and suitable for routine laboratory use. They can also withstand exposure to freezing temperatures, making them appropriate for cold storage and transport conditions. Each vial features a secure, leak resistant screw cap that can be easily opened or tightly sealed with a single turn, helping protect valuable samples from contamination or leakage. Additional features include a self standing tube design with a conical bottom for stability and molded graduations for quick and accurate volume reference during sample preparation or transfer.
Catalog No. 50-006-63188 Supplier Alkali Scientific Supplier No. C2582
Transport tubes and vials are designed for the safe storage and transport of biological materials in laboratory and research environments. Manufactured from transparent polypropylene (PP), these vials allow clear and unobstructed viewing of the sample during handling and inspection. The durable polypropylene construction provides excellent resistance to breakage, making the vials shatter proof and suitable for routine laboratory use. They can also withstand exposure to freezing temperatures, making them appropriate for cold storage and transport conditions. Each vial features a secure, leak resistant screw cap that can be easily opened or tightly sealed with a single turn, helping protect valuable samples from contamination or leakage. Additional features include a self standing tube design with a conical bottom for stability and molded graduations for quick and accurate volume reference during sample preparation or transfer.