Learn More
Med Vet International Self Tap Screws, Cruciate, J0406B, 3.5mm x 13mm, 6 Count
Catalog No. 5000582763
Encompass_Preferred
The 9/64" self-tapping screw has similar pull-out resistance to 3.5mm cortical screws. The main problem with this screw has been a lack of suitable bone plates. Our new range of 3.5mm round-hole plates considerably widens the scope of these self-tapping screws. Cruciate head. 6/pk.
Catalog No. 50-005-82763 Supplier Med Vet International Supplier No. JOR406B
The 9/64" self-tapping screw has similar pull-out resistance to 3.5mm cortical screws. The main problem with this screw has been a lack of suitable bone plates. Our new range of 3.5mm round-hole plates considerably widens the scope of these self-tapping screws. Cruciate head. 6/pk.