Learn More
Med Vet International Self Tap Screws, Cruciate, J0406N, 3.5mm x 50mm, 6 Count
Catalog No. 5000582810
Encompass_Preferred
The 9/64" self-tapping screw has similar pull-out resistance to 3.5mm cortical screws. The main problem with this screw has been a lack of suitable bone plates. Our new range of 3.5mm round-hole plates considerably widens the scope of these self-tapping screws. Cruciate head. 6/pk.
Catalog No. 50-005-82810 Supplier Med Vet International Supplier No. JOR406N
The 9/64" self-tapping screw has similar pull-out resistance to 3.5mm cortical screws. The main problem with this screw has been a lack of suitable bone plates. Our new range of 3.5mm round-hole plates considerably widens the scope of these self-tapping screws. Cruciate head. 6/pk.