Learn More
Fine Science Tools Self-Tapping Bone Screws, 100/pk
SDP
Catalog No. NC9453064
Encompass
Shaft Diameter: 1.17mm/ Alloy / Material: Stainless Steel/ Length: 4.7mm
Catalog No. NC9453064 Supplier Fine Science Tools Supplier No. 1901000
Shaft Diameter: 1.17mm/ Alloy / Material: Stainless Steel/ Length: 4.7mm