Learn More
Fine Science Tools Self-Tapping Bone Screws, 100/pk
SDP
Catalog No. NC0574761
Encompass
Shaft Diameter: 0.85mm/ Alloy / Material: Stainless Steel/ Length: 4mm
Catalog No. NC0574761 Supplier Fine Science Tools Supplier No. 1901010
Shaft Diameter: 0.85mm/ Alloy / Material: Stainless Steel/ Length: 4mm