Learn More
Med Vet International Self-Tapping Cortical Screw, J0419AS, 2.7mm x 6mm, Each
Catalog No. 5000581666
Encompass_Preferred
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Screw. Self-tapping threads. 6mm L. Stainless Steel.
Catalog No. 50-005-81666 Supplier Med Vet International Supplier No. JOR419AS
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Screw. Self-tapping threads. 6mm L. Stainless Steel.