Learn More
Med Vet International Self Tapping Screw, J0402A, 2mm x 12mm, 6 Count
Supplier: Med Vet International JOR402D
Used in conjunction with our 2mm bone plates, 2mm self-tapping screws allow the veterinary surgeon to plate the long bones of small patients or the small bones of larger patients. Until now this has been possible only with sophisticated equipment.