Learn More
Med Vet International Self Tapping Screw, J0404F, 2.7mm x 25mm, 6 Count
Catalog No. 5000582801
Encompass_Preferred
For use with 2.7mm (7/64") round-hole plates in small- to medium-sized patients. 6/pk.
Catalog No. 50-005-82801 Supplier Med Vet International Supplier No. JOR404F
For use with 2.7mm (7/64") round-hole plates in small- to medium-sized patients. 6/pk.