Learn More
Mitegen Self-Venting Funnel with Insulated Handle, For Safely Filling 5L & 10L LN2 Dewars & Dry Shippers
SDP
Catalog No. NC2084432
Encompass
This Self-Venting Funnel with Insulated Handle is an essential tool designed for the safe and efficient transfer of liquid nitrogen (LN2) into dewars and dry shippers. Its unique self-venting feature allows cryogenic gas to escape rapidly during filling, preventing splash-back and ensuring a smooth, controlled flow of LN2. The ergonomically designed insulated handle provides a secure and comfortable grip, effectively protecting users from cold burns and enhancing safety during prolonged filling procedures. This funnel is particularly useful for safely transferring liquid nitrogen into 5-liter and 10-liter dry shippers and various dewars. With approximate dimensions of an 8-3/8 inch top diameter, 8-3/4 inches in height, and a 1-inch vent diameter, it offers precise fit and optimal performance. This funnel is an indispensable accessory for any laboratory requiring reliable and safe management of cryogenic liquids.
Catalog No. NC2084432 Supplier Mitegen Supplier No. MCP111091
This Self-Venting Funnel with Insulated Handle is an essential tool designed for the safe and efficient transfer of liquid nitrogen (LN2) into dewars and dry shippers. Its unique self-venting feature allows cryogenic gas to escape rapidly during filling, preventing splash-back and ensuring a smooth, controlled flow of LN2. The ergonomically designed insulated handle provides a secure and comfortable grip, effectively protecting users from cold burns and enhancing safety during prolonged filling procedures. This funnel is particularly useful for safely transferring liquid nitrogen into 5-liter and 10-liter dry shippers and various dewars. With approximate dimensions of an 8-3/8 inch top diameter, 8-3/4 inches in height, and a 1-inch vent diameter, it offers precise fit and optimal performance. This funnel is an indispensable accessory for any laboratory requiring reliable and safe management of cryogenic liquids.