Learn More
Perkin Elmer US LLC Self-Aspirating Probe, Purple Marker, 0.25 mm I.D.
Catalog No. 502112886
Encompass_Preferred
The self-aspirating probe with orange marker includes: 0.25 mm interior diameter fluoropolymer support (180 mm) with ST nebulizer fitting 80 cm capillary.
Catalog No. 50-211-2886 Supplier Perkin Elmer US LLC Supplier No. N8152512
The self-aspirating probe with orange marker includes: 0.25 mm interior diameter fluoropolymer support (180 mm) with ST nebulizer fitting 80 cm capillary.