Learn More
Andwin Scientific SELFSEAL POUCH 5 1/2X10 200/PK
SDP
Catalog No. NC9512209
Encompass_Preferred
The 5 1/2 x 10" Convertors sterilization pouch are ideal for fast and easy presentation of sterile items. Each pouch has a chemical process indicator that inform the user of the sterilization process used. The pouches can be taped shut. This is sold in a pack of 200.
This item is not returnable. View return policy
Catalog No. NC9512209 Supplier Andwin Scientific Supplier No. 11213241
The 5 1/2 x 10" Convertors sterilization pouch are ideal for fast and easy presentation of sterile items. Each pouch has a chemical process indicator that inform the user of the sterilization process used. The pouches can be taped shut. This is sold in a pack of 200.