Learn More
Apexbio Technology LLC Roscovitine (Seliciclib,CYC202) 186692-46-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062811
Encompass_Preferred
Roscovitine (Seliciclib CYC202 CAS number 186692-46-6) is a selective inhibitor of cyclin-dependent kinases (CDKs) It targets cdc2 (CDK1/cyclin B) CDK2 (CDK2/cyclin A and CDK2/cyclin E) and CDK5 (CDK5/p35) with reported IC50 values of 0 65 M 0 7 M 0 7 M and 0 16 M respectively Additionally roscovitine inhibits extracellular signal-regulated kinases Erk1 and Erk2 displaying IC50 values of 34 M and 14 M respectively In cellular studies roscovitine has been shown to delay or halt transitions during early mitosis specifically preventing cell progression from prophase to metaphase as observed in Xenopus oocyte maturation induced by progesterone Due to its distinct inhibitory profile roscovitine is commonly utilized in research examining cell cycle regulation and kinase signaling pathways relevant to oncology
Catalog No. 50-000-62811 Supplier Apexbio Technology LLC Supplier No. A17235.1
Roscovitine (Seliciclib CYC202 CAS number 186692-46-6) is a selective inhibitor of cyclin-dependent kinases (CDKs) It targets cdc2 (CDK1/cyclin B) CDK2 (CDK2/cyclin A and CDK2/cyclin E) and CDK5 (CDK5/p35) with reported IC50 values of 0 65 M 0 7 M 0 7 M and 0 16 M respectively Additionally roscovitine inhibits extracellular signal-regulated kinases Erk1 and Erk2 displaying IC50 values of 34 M and 14 M respectively In cellular studies roscovitine has been shown to delay or halt transitions during early mitosis specifically preventing cell progression from prophase to metaphase as observed in Xenopus oocyte maturation induced by progesterone Due to its distinct inhibitory profile roscovitine is commonly utilized in research examining cell cycle regulation and kinase signaling pathways relevant to oncology