Learn More
TARGETMOL CHEMICALS INC SELINEXOR KPT-330 25MG
SDP
Catalog No. 502579556
Encompass_Preferred
Also available in 2 mg 5 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Selinexor (KPT-330) is an orally available small molecule inhibitor of CRM1 (chromosome region maintenance 1 protein exportin 1 or XPO1) with potential antineoplastic activity. purity: 99%
Catalog No. 50-257-9556 Supplier TARGETMOL CHEMICALS INC Supplier No. T610625MG
Also available in 2 mg 5 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Selinexor (KPT-330) is an orally available small molecule inhibitor of CRM1 (chromosome region maintenance 1 protein exportin 1 or XPO1) with potential antineoplastic activity. purity: 99%