Learn More
Medchemexpress LLC HY-15452 25mg , Selisistat CAS:49843-98-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1545225MG
Encompass_Preferred
HY-15452 25mg Selisistat CAS: 49843-98-3 Selisistat (EX-527) is a potent and selective SirT1 (Sir2 in Drosophila melanogaster) inhibitor with an IC50 of 123 nM for SirT1. Selisistat alleviates pathology in multiple animal and cell models of Huntington's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9749