Learn More
Medchemexpress LLC Selisistat | 49843-98-3 | C13H13ClN2O | 5 MG
SDP
Catalog No. 501873236
Encompass_Preferred
Selisistat (EX-527) is a potent and selective SirT1 (Sir2 in *Drosophila melanogaster*) inhibitor with an IC50 of 123 nM for SirT1. This compound has been shown to alleviate pathology in various animal and cell models of Huntington's disease.
  • Potent and selective SirT1 inhibitor
  • Alleviates pathology in multiple animal and cell models of Huntington's disease
  • Inhibits the deacetylation activity of human SirT1 and Drosophila Sir2 in transfected cells
  • Protective in the R6/2 mouse model of Huntington's disease
Catalog No. 50-187-3236 Supplier Medchemexpress LLC Supplier No. HY154525MG
Selisistat (EX-527) is a potent and selective SirT1 (Sir2 in *Drosophila melanogaster*) inhibitor with an IC50 of 123 nM for SirT1. This compound has been shown to alleviate pathology in various animal and cell models of Huntington's disease.
  • Potent and selective SirT1 inhibitor
  • Alleviates pathology in multiple animal and cell models of Huntington's disease
  • Inhibits the deacetylation activity of human SirT1 and Drosophila Sir2 in transfected cells
  • Protective in the R6/2 mouse model of Huntington's disease