Learn More
Selleck Chemical LLC Selisistat (EX 527) S1541-100mg
SDP
Catalog No. 503110804
Encompass_Preferred
Selisistat (EX 527 SEN0014196) is a potent and selective SIRT1 inhibitor with IC50 of 38 nM in a cell-free assay exhibits 200-fold selectivity against SIRT2 and SIRT3 Phase 2
Catalog No. 50-311-0804 Supplier Selleck Chemical LLC Supplier No. S1541100MG
Selisistat (EX 527 SEN0014196) is a potent and selective SIRT1 inhibitor with IC50 of 38 nM in a cell-free assay exhibits 200-fold selectivity against SIRT2 and SIRT3 Phase 2