Learn More
Medchemexpress LLC HY-15452B 5mg Medchemexpress, Selisistat R-enantiomer CAS:848193-69-1 Purity:>98%
SDP
Catalog No. 501932740
Encompass_Preferred
Medchemexpress, HY-15452B 5mg Selisistat R-enantiomer CAS:848193-69-1 Selisistat R-enantiomer (EX-527 R-enantiomer) is much less active R-enantiomer of Selisistat, with an IC 50 of > 100 μM for SIRT1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2740 Supplier Medchemexpress LLC Supplier No. HY15452B5MG
Medchemexpress, HY-15452B 5mg Selisistat R-enantiomer CAS:848193-69-1 Selisistat R-enantiomer (EX-527 R-enantiomer) is much less active R-enantiomer of Selisistat, with an IC 50 of > 100 μM for SIRT1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.