Learn More
Medchemexpress LLC HY-15452A 5mg Medchemexpress, Selisistat S-enantiomer CAS:848193-68-0 Purity:>98%
SDP
Catalog No. 501932739
Encompass_Preferred
Medchemexpress, HY-15452A 5mg Selisistat S-enantiomer CAS:848193-68-0 Selisistat S-enantiomer (EX-527 S-enantiomer) is the S-enantiomer of Selisistat, with an IC 50 of 123 nM for SIRT1. Selisistat S-enantiomer is much more potent than Selisistat R-enantiomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2739 Supplier Medchemexpress LLC Supplier No. HY15452A5MG
Medchemexpress, HY-15452A 5mg Selisistat S-enantiomer CAS:848193-68-0 Selisistat S-enantiomer (EX-527 S-enantiomer) is the S-enantiomer of Selisistat, with an IC 50 of 123 nM for SIRT1. Selisistat S-enantiomer is much more potent than Selisistat R-enantiomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.