Learn More
Aviva Systems Biology Corp SELL Antibody - C-terminal region (ARP63028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SELL. It was validated on Western Blot. . Alternate names: CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-287-848