Learn More
Selleck Chemical LLC Docetaxel 50mg 114977-28-5 RP56976, NSC 628503
SDP
Supplier:  Selleck Chemical LLC S114850MG
Encompass_Preferred
Docetaxel (RP56976, NSC 628503), an analog of paclitaxel, is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1139264