Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Sellstrom Face Shield, Clear, 8 x 0.04 in, 1 ea

Catalog No. 5000796174
Encompass

This product is a clear face shield window designed for personal protective equipment, providing essential eye and face protection during various tasks. Its primary purpose is to serve as a replaceable component for a face shield system, ensuring clear visibility while offering a barrier against hazards.

  • Material: Polycarbonate
  • Coating: Bright/uncoated
  • Lens shade: None
  • Window height: 8 in
  • Window width: 16 in
  • Window thickness: 0.04 in
  • Certifications: Meets CAN/CSA Z94.3 and ANSI Z87.1+ standards
  • Compatibility: Designed for use with a specific series of crown headgear

Catalog No. 50-007-96174 Supplier MSC Supplier No. 00370643
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a clear face shield window designed for personal protective equipment, providing essential eye and face protection during various tasks. Its primary purpose is to serve as a replaceable component for a face shield system, ensuring clear visibility while offering a barrier against hazards.

  • Material: Polycarbonate
  • Coating: Bright/uncoated
  • Lens shade: None
  • Window height: 8 in
  • Window width: 16 in
  • Window thickness: 0.04 in
  • Certifications: Meets CAN/CSA Z94.3 and ANSI Z87.1+ standards
  • Compatibility: Designed for use with a specific series of crown headgear

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.