Learn More
MSC Sellstrom Face Shield and Headgear, Polycarbonate, Anti-Fog, 1 ea
Catalog No. 5000781762
Encompass
This face shield and headgear set features a polycarbonate window and nylon headgear. It provides protection with a clear, anti-fog window and an adjustable ratchet-style headgear.
  • Anti-fog window coating
  • Polycarbonate window material
  • Black nylon headgear
  • Clear window color
  • Window dimensions: 9 in high, 12-1/8 in wide
  • Window thickness: 0.0600 in
  • Ratchet headgear adjustment
  • Molded and stylish crown features
  • Meets CE and CSA Z94.3-2015 standards
  • Compatible with ANSI Type I, Class E hard hats
Catalog No. 50-007-81762 Supplier MSC Supplier No. 44953396
This face shield and headgear set features a polycarbonate window and nylon headgear. It provides protection with a clear, anti-fog window and an adjustable ratchet-style headgear.
  • Anti-fog window coating
  • Polycarbonate window material
  • Black nylon headgear
  • Clear window color
  • Window dimensions: 9 in high, 12-1/8 in wide
  • Window thickness: 0.0600 in
  • Ratchet headgear adjustment
  • Molded and stylish crown features
  • Meets CE and CSA Z94.3-2015 standards
  • Compatible with ANSI Type I, Class E hard hats