Learn More
MSC Sellstrom Face Shield Headgear, Polycarbonate, Black, DP4 Series, 1 ea
Catalog No. 5000794933
Encompass
This product is a replacement face shield headgear from Sellstrom. It is designed to offer robust protection and comfort, suitable for various industrial applications requiring face protection.
  • Accessory type: face shield headgear replacement
  • Material: polycarbonate
  • Color: black
  • Product compatibility: DP4 series headgear
  • Country of origin: Taiwan
Catalog No. 50-007-94933 Supplier MSC Supplier No. 39988829
This product is a replacement face shield headgear from Sellstrom. It is designed to offer robust protection and comfort, suitable for various industrial applications requiring face protection.
  • Accessory type: face shield headgear replacement
  • Material: polycarbonate
  • Color: black
  • Product compatibility: DP4 series headgear
  • Country of origin: Taiwan