Learn More
MSC Sellstrom Goggles, Odyssey II, Anti-Fog, Clear Polycarbonate, 1 ea
Catalog No. 5000795674
Encompass
Sellstrom Odyssey II Industrial Goggles are designed for superior protection against chemical splash and dust. Featuring a dual-lens design with anti-fog and anti-scratch coatings, these goggles ensure clear vision in various industrial environments.
  • Indirect vented body allows air ventilation while protecting against liquids and dust
  • Dual UV absorbing polycarbonate lens
  • Outer lens is anti-scratch and high impact resistant
  • Inner lens has an anti-fog clear coating
  • Flexible goggle ensures proper fit
  • Can be used comfortably over most prescription glasses
  • Soft body design conforms to face for a comfortable seal
  • Exceptional panoramic view
  • Meets ANSI Z87.1+ high impact standards
  • Includes elastic strap
Catalog No. 50-007-95674 Supplier MSC Supplier No. 41854779
Sellstrom Odyssey II Industrial Goggles are designed for superior protection against chemical splash and dust. Featuring a dual-lens design with anti-fog and anti-scratch coatings, these goggles ensure clear vision in various industrial environments.
  • Indirect vented body allows air ventilation while protecting against liquids and dust
  • Dual UV absorbing polycarbonate lens
  • Outer lens is anti-scratch and high impact resistant
  • Inner lens has an anti-fog clear coating
  • Flexible goggle ensures proper fit
  • Can be used comfortably over most prescription glasses
  • Soft body design conforms to face for a comfortable seal
  • Exceptional panoramic view
  • Meets ANSI Z87.1+ high impact standards
  • Includes elastic strap