Learn More
Sellstrom™ Guest-Gard™ Glasses
Catalog No. 17287A
Click to view available options
Clear lens tint
Catalog No.
17-287A Clear lens tint
1 options
This item is not returnable. View return policy
Catalog No. 17-287A Supplier Sellstrom™ Supplier No. S79103
This item is not returnable. View return policy
Lightweight, comfortable glasses can be worn over prescription glasses
  • Universal fit nose bridge
  • Fixed sideshields
  • Frosted browguard
  • 146mm temples
Order Info
Pack of 24
Compliance
Meet ANSI impact standards
Specifications
Certifications/Compliance Meets ANSI Z87.1-2015 standards
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Visitor's spec; Everyday eye protection
Frame Color Clear
Quantity 24/dispenser box