Learn More
Sellstrom™ Model 2000 Monitor Germicidal Cabinet
Catalog No. 179365
Click to view available options
Product Type:
Germicidal Cabinet
Catalog No. Product Type
17-936-5 Germicidal Cabinet
1 options
Catalog No. 17-936-5 Supplier Sellstrom™ Supplier No. S90494
The ultimate value in germicidal eyewear monitors, the Model 2000, featuring Hyper-Germicidal™ quartz bulb with long-lasting ballast, is designed for 20 years of service. Other features include heavy-duty five-minute timer, two tough key locks with vault rods, and 24-gauge white enamel steel construction.
Independent testing confirms that the Model 2000 kills 99.2% of bacteria in a five-minute exposure. Double doors seal in UV radiation. Pilot light indicates when UV lamp is on. Automatic shutoff safely turns off bulb if doors are accidentally opened during UV cycle. Holds six glasses or five goggles per shelf, eight shelves per cabinet. Wall or shelf mountable with included 7 ft. (2.1m) grounded electrical cord. Space-saving dimensions: 9 1/2D × 24 1/2W × 32 in.H (24.1 × 62.2 × 81.2cm). Weight: 45 lb. (99kg). UL listed.
Specifications
Description Shop Model
Product Type Germicidal Cabinet